missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethoxymethyl Chloride, TRC

Product Code. 30452059
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452059 Supplier Toronto Research Chemicals Supplier No. TRCT7876001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Trimethylsilyl)ethoxymethyl Chloride
CAS 76513-69-4
Molecular Formula C6 H15 Cl O Si
Quantity 1 g
Synonym 2-[(Chloromethoxy)ethyl]trimethylsilane, 2-(Trimethylsilanyl)ethoxymethyl Chloride, Chloromethyl 2-(Trimethylsilyl)ethyl Ether, Chloromethyl Trimethylsilylethyl Ether, SEM-Cl, SEM Cl, SEM Chloride, [2-[(Chloromethyl)oxy]ethyl]trimethylsilane, [[2-(Trimethylsilyl)ethyl]oxy]methyl Chloride, β-(Trimethylsilyl)ethoxymethyl Chloride
InChI Formula InChI=1S/C6H15ClOSi/c1-9(2,3)5-4-8-6-7/h4-6H2,1-3H3
SMILES C[Si](C)(C)CCOCCl
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
Molecular Weight (g/mol) 166.72
Formula Weight 166.0581
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.