missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized

Product Code. 10442221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442221 5 mL Glass bottle 5mL
10712934 25 mL Glass bottle 25mL
11975821 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10442221 Supplier Thermo Scientific Acros Supplier No. 219020050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76513-69-4 | C6H15ClOSi | 166.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76513-69-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
InChI Key BPXKZEMBEZGUAH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
PubChem CID 2724271
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
SMILES C[Si](C)(C)CCOCCl

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 57.0°C to 59.0°C (8.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2O(CH2)2Si(CH3)3
Refractive Index 1.4350 to 1.4370
Quantity 5 mL
Fieser 10,431; 16,361; 01,107
Specific Gravity 0.94
Solubility Information Solubility in water: decomposes
Stabilizer 10ppm diisopropylethylamine
Formula Weight 166.72
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)ethoxymethyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trimethylsilyl)ethoxymethyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.