missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethanol, 98+%, Thermo Scientific Chemicals

Catalog Number 11460037
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2916-68-9
C5H14OSi
118.251
MFCD00002825
ZNGINKJHQQQORD-UHFFFAOYSA-N
2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
18013
2-trimethylsilylethanol
C[Si](C)(C)CCO
This item is not returnable. View return policy
2916-68-9
C5H14OSi
118.251
MFCD00002825
ZNGINKJHQQQORD-UHFFFAOYSA-N
2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
18013
2-trimethylsilylethanol
C[Si](C)(C)CCO

2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Density 0.825
Boiling Point 71°C to 72°C (10 mm)
Flash Point 50°C (122°F)
Assay Percent Range 98+%
Refractive Index 1.425
Quantity 5 g
UN Number UN1987
Beilstein 1732034
Solubility Information Miscible with water.
Formula Weight 118.25
Percent Purity ≥98%
Chemical Name or Material 2-(Trimethylsilyl)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'