missing translation for 'onlineSavingsMsg'
Learn More

2-Trimethylsilyl-1-ethylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11328916
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328916 250 mg 250mg
11318916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328916 Supplier Thermo Scientific Alfa Aesar Supplier No. L19534.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165904-20-1
Molecular Formula C11H25BO2Si
Molecular Weight (g/mol) 228.21
MDL Number MFCD03788725
InChI Key YKYAZAJUMAWTGO-UHFFFAOYSA-N
Synonym 2-trimethylsilyl-1-ethylboronic acid pinacol ester, trimethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl silane, trimethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl silane
PubChem CID 11436111
IUPAC Name trimethyl-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]silane
SMILES CC1(C)OB(CC[Si](C)(C)C)OC1(C)C

Specifications

Refractive Index 1.431
Quantity 250 mg
Beilstein 8683473
Sensitivity Moisture sensitive
Formula Weight 228.22
Percent Purity 97%
Chemical Name or Material 2-Trimethylsilyl-1-ethylboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.