missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsiloxy)ethyl methacrylate, 94%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 0.995 |
| Boiling Point | 65°C (0.9 mmHg) |
| Flash Point | 42°C (107°F) |
| Refractive Index | 1.43 |
| Quantity | 50 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Difficult to mix. |
| Formula Weight | 202.33 |
| Percent Purity | 94% |
| Chemical Name or Material | 2-(Trimethylsiloxy)ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction