missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)quinoxaline, 97%, Thermo Scientific™

Product Code. 10744312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10744312 5 g Glass bottle 5g
10441440 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10744312 Supplier Thermo Scientific Acros Supplier No. 432630050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 148853-42-3
Molecular Formula C9H5F3N2
Molecular Weight (g/mol) 198.15
MDL Number MFCD06656411
InChI Key YIQVNKFWTSLVSA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl quinoxaline, quinoxaline,2-trifluoromethyl, trifluoromethyl quinoxaline, acmc-1c899
PubChem CID 10352693
IUPAC Name 2-(trifluoromethyl)quinoxaline
SMILES FC(F)(F)C1=NC2=CC=CC=C2N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 65°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 198.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)quinoxaline
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)quinoxaline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.