missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%

Product Code. 15490746
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490746 250 mg 250mg
15470746 1 g 1g
15480746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490746 Supplier Thermo Scientific Alfa Aesar Supplier No. H54088.MD
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 306960-77-0 | C6H3F3N2O2 | 192.10 g/mol

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306960-77-0
Molecular Formula C6H3F3N2O2
Molecular Weight (g/mol) 192.10
MDL Number MFCD09054851
InChI Key RGLZXVNOHDUMRJ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidine-5-carboxylic acid, 2-trifluoromethyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-trifluoromethyl, 5-pyrimidinecarboxylic acid, 2-trifluoromethyl-9ci, pubchem13190, 2-trifluoromethylpyrimidine-5-carboxylicacid, 2-trifluoromethyl pyrimidine-5-carboxylicacid, 2-trifluoromethyl-5-pyrimidinecarboxilic acid, 2-trifluoromethyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-trifluoromethyl
PubChem CID 10583889
IUPAC Name 2-(trifluoromethyl)pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=C(N=C1)C(F)(F)F

Specifications

Melting Point 170°C to 175°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 192.1
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.