missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals

Product Code. 15499557
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499557 1 g 1g
15470126 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499557 Supplier Thermo Scientific Alfa Aesar Supplier No. H52803.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914348-10-0
Molecular Formula C6H4F3N3O
Molecular Weight (g/mol) 191.113
MDL Number MFCD05662689
InChI Key LBNILUATVOCQRF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidine-4-carboxamide, 4-carbamoyl-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-4-pyrimidinecarboxamide, 2-trifluoromethyl-pyrimidine-4-carboxylic acid amide, 2-trifluoromethylpyrimidine-4-carboxylic acid amide
PubChem CID 17750126
IUPAC Name 2-(trifluoromethyl)pyrimidine-4-carboxamide
SMILES C1=CN=C(N=C1C(=O)N)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrimidine-4-carboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.