missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals

Catalog Number 15470126
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
914348-10-0
C6H4F3N3O
191.113
MFCD05662689
LBNILUATVOCQRF-UHFFFAOYSA-N
2-trifluoromethyl pyrimidine-4-carboxamide, 4-carbamoyl-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-4-pyrimidinecarboxamide, 2-trifluoromethyl-pyrimidine-4-carboxylic acid amide, 2-trifluoromethylpyrimidine-4-carboxylic acid amide
17750126
2-(trifluoromethyl)pyrimidine-4-carboxamide
C1=CN=C(N=C1C(=O)N)C(F)(F)F
This item is not returnable. View return policy
914348-10-0
C6H4F3N3O
191.113
MFCD05662689
LBNILUATVOCQRF-UHFFFAOYSA-N
2-trifluoromethyl pyrimidine-4-carboxamide, 4-carbamoyl-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-4-pyrimidinecarboxamide, 2-trifluoromethyl-pyrimidine-4-carboxylic acid amide, 2-trifluoromethylpyrimidine-4-carboxylic acid amide
17750126
2-(trifluoromethyl)pyrimidine-4-carboxamide
C1=CN=C(N=C1C(=O)N)C(F)(F)F

2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrimidine-4-carboxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'