missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals

Catalog Number 11388946
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
368-48-9
C6H4F3N
147.10
MFCD02183562
ATRQECRSCHYSNP-UHFFFAOYSA-N
2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
136199
2-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=CC=N1
This item is not returnable. View return policy
368-48-9
C6H4F3N
147.10
MFCD02183562
ATRQECRSCHYSNP-UHFFFAOYSA-N
2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
136199
2-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=CC=N1

Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.

Solubility
Soluble in most organic solvents.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.28
Boiling Point 139°C to 141°C
Flash Point 42°C (108°F)
Refractive Index 1.418
Quantity 1 g
UN Number UN1993
Beilstein 3112
Solubility Information Soluble in most organic solvents.
Formula Weight 147.1
Percent Purity 99%
Chemical Name or Material 2-(Trifluoromethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'