missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylhydrazine, 97%

Product Code. 11489120
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489120 1 g 1g
11499120 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489120 Supplier Thermo Scientific Alfa Aesar Supplier No. A14781.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 365-34-4 | C7H7F3N2 | 176.142 g/mol

2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 365-34-4
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00052687
InChI Key JSWQDLBFVSTSIW-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylhydrazine, 2-trifluoromethyl phenyl hydrazine, 2-trifluoromethylphenylhydrazine, 1-2-trifluoromethyl phenyl hydrazine, m-trifluoromethylphenylhydrazine, hydrazine, 2-trifluoromethyl phenyl, buttpark 44\57-17, 2-trifluoromethyl-phenyl-hydrazine
PubChem CID 520800
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN

Specifications

Melting Point 57°C to 63°C
Quantity 1 g
Beilstein 641592
Solubility Information Insoluble in water.
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.