missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

Product Code. 11329156
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329156 250 mg 250mg
11309156 1 g 1g
11319156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329156 Supplier Thermo Scientific Alfa Aesar Supplier No. L20280.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 745783-91-9 | C9H5F3O2 | 202.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745783-91-9
Molecular Formula C9H5F3O2
Molecular Weight (g/mol) 202.13
MDL Number MFCD08533288
InChI Key HBOSPPHIPGYHDI-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylglyoxal hydrate, 2-oxo-2-2-trifluoromethyl phenyl acetaldehyde hydrate, 2-oxo-2-2-trifluoromethyl phenyl ethanal, oxamethane, benzeneacetaldehyde, a-oxo-2-trifluoromethyl-,hydrate 1:1
PubChem CID 2783281
IUPAC Name 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde;hydrate
SMILES FC(F)(F)C1=CC=CC=C1C(=O)C=O

Specifications

Melting Point 36°C to 43°C
Quantity 250 mg
Formula Weight 202.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 2-(Trifluoromethyl)phenylglyoxal hydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.