missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylacetylene, 97%

Product Code. 15450338
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450338 5 g 5g
15440338 1 g 1g
15460338 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450338 Supplier Thermo Scientific Alfa Aesar Supplier No. H53478.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 704-41-6 | C9H5F3 | 170.134 g/mol

2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-41-6
Molecular Formula C9H5F3
Molecular Weight (g/mol) 170.134
MDL Number MFCD00467313
InChI Key OOZKONVIIMFOKW-UHFFFAOYSA-N
Synonym 1-ethynyl-2-trifluoromethyl benzene, 2'-trifluoromethylphenyl acetylene, 2-trifluoromethyl phenylacetylene, 2-ethynyl-alpha,alpha,alpha-trifluorotoluene, 2-ethynyl-a,a,a-trifluorotoluene,1-ethynyl-2-trifluoromethylbenzene, 2'-trifluoromethylphenylacetylene, 2-ethynylbenzotrifluoride, o-ethynyl-trifluoromethylbenzene, 2-trifluoromethyl phenyl ethyne, 1-ethynyl-2-trifluoromethyl-benzene
PubChem CID 4100856
IUPAC Name 1-ethynyl-2-(trifluoromethyl)benzene
SMILES C#CC1=CC=CC=C1C(F)(F)F

Specifications

Density 1.222
Boiling Point 45°C (8 mmHg)
Flash Point 25°C (77°F)
Refractive Index 1.476
Quantity 5 g
UN Number UN1993
Solubility Information Sparingly soluble in water.
Formula Weight 170.13
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.