missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenothiazine, TRC

Product Code. 30453380
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. Quantity unitSize
30453380 1 g 1g
30449715 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30453380

Brand: Toronto Research Chemicals TRCT7913701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-30-8
Formula Weight 267.033
InChI Formula InChI=1S/C13H8F3NS/c14-13(15,16)8-5-6-12-10(7-8)17-9-3-1-2-4-11(9)18-12/h1-7,17H
IUPAC Name 2-(trifluoromethyl)-10H-phenothiazine
Molecular Formula C13H8F3NS
Molecular Weight (g/mol) 267.27
SMILES FC(F)(F)C1=CC(N2)=C(C=C1)SC3=C2C=CC=C3
Synonym 2-(Trifluoromethyl)-10H-phenothiazine, 2-(Trifluoromethyl)phenothiazine, NSC 50438
Chemical Name or Material 2-(Trifluoromethyl)phenothiazine

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.