missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenol, 98%

Product Code. 11412347
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412347 25 g 25g
11399599 1 g 1g
11402347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412347 Supplier Thermo Scientific Alfa Aesar Supplier No. A14231.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-30-4
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002222
InChI Key ZOQOPXVJANRGJZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
PubChem CID 67958
IUPAC Name 2-(trifluoromethyl)phenol
SMILES C1=CC=C(C(=C1)C(F)(F)F)O

Specifications

Melting Point 45°C to 46°C
Boiling Point 147°C to 148°C
Flash Point 65°C (149°F)
Odor Phenol-like
Quantity 25 g
UN Number UN3261
Beilstein 1867917
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.