missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)oxazolo[4,5-b]pyridine, 97%, Thermo Scientific™

Product Code. 15450909
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15450909 5 g 5g
15458586 250 mg 250mg
15440909 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15450909

Brand: Thermo Scientific Alfa Aesar H63931.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 894406-63-4
Molecular Formula C7H3F3N2O
Molecular Weight (g/mol) 188.109
InChI Key MJILPMZYOJELAX-UHFFFAOYSA-N
PubChem CID 29919200
IUPAC Name 2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
SMILES C1=CC2=C(N=C1)N=C(O2)C(F)(F)F

Specifications

Quantity 5 g
Synonym 2-trifluoromethyl oxazolo 4,5-b pyridine, 2-trifluoromethyl-1,3 oxazolo 4,5-b pyridine, oxazolo 4,5-b pyridine,2-trifluoromethyl, oxazolo 4,5-b pyridine, 2-trifluoromethyl, 2-trifluoromethyl 1,3 oxazolo 4,5-b pyridine
Formula Weight 188.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)oxazolo[4,5-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.