missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 15454487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454487 250 mg 250mg
15444487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454487 Supplier Thermo Scientific Alfa Aesar Supplier No. H31841.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-43-8
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD01862052
InChI Key BFROETNLEIAWNO-UHFFFAOYSA-N
Synonym 2-trifluoromethyl nicotinic acid, 2-trifluoromethylnicotinic acid, 2-trifluoromethyl pyridine-3-carboxylic acid, 2-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-3-pyridinecarboxylic acid, pubchem12728, acmc-1c9gb, trifluoromethylnicotinic acid
PubChem CID 2777547
IUPAC Name 2-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(N=CC=C1)C(F)(F)F

Specifications

Melting Point 185°C to 187°C
Quantity 250 mg
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.