missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzylamine, 98%

Product Code. p-205078 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10402845

Brand: Thermo Scientific Acros 312950010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3048-01-9 | C8H8F3N | 175.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3048-01-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.16
MDL Number MFCD00010297
InChI Key ZSKQIFWUTUZAGF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzylamine, 2-trifluoromethylbenzylamine, 2-trifluoromethyl phenyl methanamine, o-trifluoromethylbenzyl amine, o-trifluoromethylbenzylamine, 2-trifluoromethyl-benzylamine, 1-2-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-trifluoromethyl, o-trifluoromethyl benzylamine
PubChem CID 76447
IUPAC Name [2-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2490g/mL
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CF3C6H4CH2NH2
Refractive Index 1.4700 to 1.472
Quantity 1 g
Specific Gravity 1.249
Formula Weight 175.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzylamine, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.