missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoyl Chloride, TRC

Product Code. 30452330
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30452330 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30452330

Brand: Toronto Research Chemicals TRCT791135100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Trifluoromethyl)benzoyl Chloride
CAS 312-94-7
Molecular Formula C8H4ClF3O
Quantity 100 g
Synonym alpha,alpha,alpha-Trifluoro-o-toluoyl Chloride, NSC 88290, o-(Trifluoromethyl)benzoyl Chloride
InChI Formula InChI=1S/C8H4ClF3O/c9-7(13)5-3-1-2-4-6(5)8(10,11)12/h1-4H
SMILES FC(F)(F)c1ccccc1C(=O)Cl
IUPAC Name 2-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol) 208.56
Formula Weight 207.9903
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.