missing translation for 'onlineSavingsMsg'
Learn More

(2-Trifluoromethyl-benzoimidazol-1-yl)-acetic acid, ≥97%, Thermo Scientific™

Product Code. 10653425
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10653425 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10653425 Supplier Thermo Scientific Maybridge Supplier No. MO07742DA
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 313241-14-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H7F3N2O2
MDL Number MFCD00994711
Quantity 1 g
Synonym 2-trifluoromethyl-benzoimidazol-1-yl-acetic acid, 2-trifluoromethyl-1h-benzimidazol-1-yl acetic acid, 1h-benzimidazole-1-aceticacid, 2-trifluoromethyl, 2-trifluoromethyl-1h-benzimidazol-1-yl-acetic acid, 2-2-trifluoromethyl-1h-1,3-benzodiazol-1-yl acetic acid, 2-2-trifluoromethyl benzimidazolyl acetic acid, 2-2-trifluoromethyl-1h-benzo d imidazol-1-yl acetic acid, 2-2-trifluoromethyl-1h-1,3-benzimidazol-1-yl acetic acid, 2-trifluoromethyl-1,3-benzodiazol-1-yl acetic acid
InChI Key GPOYZFXQCCJBTF-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)N=C(N2CC(=O)O)C(F)(F)F
IUPAC Name 2-[2-(trifluoromethyl)benzimidazol-1-yl]acetic acid
Molecular Weight (g/mol) 244.173
PubChem CID 675083
Formula Weight 244.17
Percent Purity 97%
Chemical Name or Material [2-(Trifluoromethyl)-1H-benzimidazol-1-yl]acetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.