missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)benzoic Acid, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 2-(Trifluoromethyl)benzoic acid |
| CAS | 433-97-6 |
| Molecular Formula | C8 H5 F3 O2 |
| Quantity | 100 g |
| Synonym | o-Toluic acid','α,α,α-trifluoro- (6CI,7CI,8CI), 2-(Trifluoromethyl)benzoic acid, NSC 88326, o-(Trifluoromethyl)benzoic acid, α,α,α-Trifluoro-o-toluic acid |
| InChI Formula | InChI=1S/C8H5F3O2/c9-8(10,11)6-4-2-1-3-5(6)7(12)13/h1-4H,(H,12,13) |
| SMILES | OC(=O)c1ccccc1C(F)(F)F |
| IUPAC Name | 2-(trifluoromethyl)benzoic acid |
| Molecular Weight (g/mol) | 190.12 |
| Formula Weight | 190.0242 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction