missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15535056 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535056 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535056 Supplier Thermo Scientific Acros Supplier No. 188451000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 433-97-6
Melting Point 107.0°C to 110.0°C
Boiling Point 247.0°C (753.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
MDL Number MFCD00002476
Quantity 100 g
Beilstein 09,III,2308
Synonym 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid
InChI Key FBRJYBGLCHWYOE-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1C(F)(F)F
IUPAC Name 2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 190.12
PubChem CID 9899
ChEBI CHEBI:60694
Formula Weight 190.12
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Trifluoromethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.