missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10512981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10512981 5 g Glass bottle 5g
10742125 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10512981 Supplier Thermo Scientific Acros Supplier No. 188450050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-97-6
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002476
InChI Key FBRJYBGLCHWYOE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid
PubChem CID 9899
ChEBI CHEBI:60694
IUPAC Name 2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 107.0°C to 110.0°C
Color Brown-Yellow to Yellow
Boiling Point 247.0°C (753.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3C6H4CO2H
Quantity 5 g
Beilstein 09, III, 2308
Formula Weight 190.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Trifluoromethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.