Learn More
2-(Trifluoromethyl)benzenethiol, ≥95%, Thermo Scientific™
£12.75 - £94.35
Chemical Identifiers
CAS | 13333-97-6 |
---|---|
Molecular Formula | C7H5F3S |
Molecular Weight (g/mol) | 178.172 |
MDL Number | MFCD00068896 |
InChI Key | XVGQHRKNXSUPEF-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol |
PubChem CID | 2777889 |
IUPAC Name | 2-(trifluoromethyl)benzenethiol |
SMILES | C1=CC=C(C(=C1)C(F)(F)F)S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10079541
|
Thermo Scientific Maybridge
TL00684DA |
1 g | Amber Glass Bottle |
£12.75
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10520173
|
Thermo Scientific Maybridge
TL00684EA |
10 g | Amber Glass Bottle |
£94.35
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
13333-97-6 | |
178.172 | |
XVGQHRKNXSUPEF-UHFFFAOYSA-N | |
2777889 | |
C1=CC=C(C(=C1)C(F)(F)F)S |
C7H5F3S | |
MFCD00068896 | |
2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol | |
2-(trifluoromethyl)benzenethiol |
Specifications
13333-97-6 | |
1.35g/mL | |
C7H5F3S | |
MFCD00068896 | |
XVGQHRKNXSUPEF-UHFFFAOYSA-N | |
2-(trifluoromethyl)benzenethiol | |
2777889 | |
95% |
95.0 | |
95% | |
CF3C6H4SH | |
2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol | |
C1=CC=C(C(=C1)C(F)(F)F)S | |
178.172 | |
178.18 | |
2-(Trifluoromethyl)benzenethiol |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning