missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzamide, 98+%, Thermo Scientific™

Product Code. 11438056
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438056 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438056 Supplier Thermo Scientific Alfa Aesar Supplier No. A14103.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 360-64-5
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD00014798
InChI Key QBAYIBZITZBSFO-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzamide, o-trifluoromethylbenzamide, o-trifluoromethyl benzamide, unii-5u8y7e6aio, 2-trifluoromehtyl benzamide, benzamide, 2-trifluoromethyl, o-trifluoromethyl-benzamide, 5u8y7e6aio, synquest 4848-3-12
PubChem CID 67759
IUPAC Name 2-(trifluoromethyl)benzamide
SMILES NC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 161°C to 164°C
Quantity 25 g
Beilstein 2616148
Formula Weight 189.14
Percent Purity ≥98%
Chemical Name or Material 2-(Trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.