missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzaldehyde, 98%

Product Code. 11450438
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450438 5 g 5g
11460438 25 g 25g
11470438 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450438 Supplier Thermo Scientific Alfa Aesar Supplier No. B24591.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 447-61-0 | C8H5F3O | 174.122 g/mol

2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

Solubility
Soluble in most organic solvents.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 447-61-0
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00003337
InChI Key ZDVRPKUWYQVVDX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzaldehyde, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, 2-trifluoromethyl-benzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde
PubChem CID 123067
IUPAC Name 2-(trifluoromethyl)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)C(F)(F)F

Specifications

Melting Point -40°C
Density 1.322
Boiling Point 70°C to 71°C (16 mmHg)
Flash Point 61°C (141°F)
Odor Almond-like
Refractive Index 1.466
Quantity 5 g
Beilstein 2045512
Sensitivity Air Sensitive
Solubility Information Soluble in most organic solvents.
Formula Weight 174.12
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.