missing translation for 'onlineSavingsMsg'
Learn More

2'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™

Product Code. 10685852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10685852 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10685852 Supplier Thermo Scientific Acros Supplier No. 214430010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17408-14-9
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2500g/mL
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Refractive Index 1.4584 to 1.4604
MDL Number MFCD00000378
Quantity 1 g
Synonym 2'-trifluoromethyl acetophenone, 1-2-trifluoromethyl phenyl ethanone, o-trifluoromethylacetophenone, 1-2-trifluoromethyl phenyl ethan-1-one, 2'-trifluoromethylacetophenone, ethanone, 1-2-trifluoromethyl phenyl, o-trifluoromethyl benzoyl, 2-trifluoromethylacetophenone, 1-acetyl-2-trifluoromethyl benzene
InChI Key FYDUUODXZQITBF-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC=C1C(F)(F)F
IUPAC Name 1-[2-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol) 188.149
PubChem CID 87095
Formula Weight 188.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2′-(Trifluoromethyl)acetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-(Trifluoromethyl)acetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.