missing translation for 'onlineSavingsMsg'
Learn More

2'-(Trifluoromethyl)acetophenone, 97+%, Thermo Scientific™

Product Code. 11414953
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414953 5 g 5g
11424953 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414953 Supplier Thermo Scientific Alfa Aesar Supplier No. A12248.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17408-14-9
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000378
InChI Key FYDUUODXZQITBF-UHFFFAOYSA-N
Synonym 2'-trifluoromethyl acetophenone, 1-2-trifluoromethyl phenyl ethanone, o-trifluoromethylacetophenone, 1-2-trifluoromethyl phenyl ethan-1-one, 2'-trifluoromethylacetophenone, ethanone, 1-2-trifluoromethyl phenyl, o-trifluoromethyl benzoyl, 2-trifluoromethylacetophenone, 1-acetyl-2-trifluoromethyl benzene
PubChem CID 87095
IUPAC Name 1-[2-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 15°C to 17°C
Density 1.255
Boiling Point 163°C to 165°C
Flash Point 84°C (183°F)
Odor Characteristic
Refractive Index 1.4595
Quantity 5 g
Beilstein 2259035
Formula Weight 188.15
Percent Purity ≥97%
Chemical Name or Material 2'- (Trifluoromethyl)acetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.