missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(trifluoromethoxy)phenyl isothiocyanate, Thermo Scientific™
£15.00 - £33.40
Chemical Identifiers
CAS | 175205-33-1 |
---|---|
Molecular Formula | C8H4F3NOS |
Molecular Weight (g/mol) | 219.18 |
MDL Number | MFCD00220860 |
InChI Key | ZFEJYPIWOGCCFS-UHFFFAOYSA-N |
PubChem CID | 2777336 |
IUPAC Name | 1-isothiocyanato-2-(trifluoromethoxy)benzene |
SMILES | FC(F)(F)OC1=CC=CC=C1N=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10119382
|
Thermo Scientific Maybridge
SB01674EA |
10 g |
£15.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10285594
|
Thermo Scientific Maybridge
SB01674EB |
25 g |
£33.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
175205-33-1 | |
219.18 | |
ZFEJYPIWOGCCFS-UHFFFAOYSA-N | |
1-isothiocyanato-2-(trifluoromethoxy)benzene |
C8H4F3NOS | |
MFCD00220860 | |
2777336 | |
FC(F)(F)OC1=CC=CC=C1N=C=S |
Specifications
175205-33-1 | |
C8H4F3NOS | |
2-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethoxy benzene, benzene,1-isothiocyanato-2-trifluoromethoxy, 1-isothiocyanato-2-trifluoromethoxy-benzene, 2-trifluoromethoxy phenylisothiocyanate, benzene, 1-isothiocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisothiocyanate, 1-isothiocyanato-2-trifluoromethyloxy benzene, 2-isothiocyanato-alpha,alpha,alpha-trifluoroanisole | |
FC(F)(F)OC1=CC=CC=C1N=C=S | |
219.18 | |
219.19 | |
2-(Trifluoromethoxy)phenyl isothiocyanate |
97% | |
MFCD00220860 | |
ZFEJYPIWOGCCFS-UHFFFAOYSA-N | |
1-isothiocyanato-2-(trifluoromethoxy)benzene | |
2777336 | |
97% |
Safety and Handling
GHS Signal Word: Warning