missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)phenol, 97%, Thermo Scientific™

Product Code. 11444287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444287 5 g 5g
11434287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444287 Supplier Thermo Scientific Alfa Aesar Supplier No. B20376.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32858-93-8
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00236324
InChI Key GQWMNVOVQZIPJC-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
PubChem CID 2777299
IUPAC Name 2-(trifluoromethoxy)phenol
SMILES C1=CC=C(C(=C1)O)OC(F)(F)F

Specifications

Density 1.332
Boiling Point 69°C to 71°C (60 mmHg)
Refractive Index 1.443
Quantity 5 g
Beilstein 1869013
Formula Weight 178.11
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.