missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzenesulfonamide, TRC

Product Code. 30451361
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451361 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451361 Supplier Toronto Research Chemicals Supplier No. TRCT7971582.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2-(Trifluoromethoxy)benzenesulfonamide
CAS 37526-59-3
Molecular Formula C7H6NO3F3S
Quantity 2.5 g
Synonym 2-(Trifluoromethoxy)benzene-1-sulfonamide
InChI Formula InChI=1S/C7H6F3NO3S/c8-7(9,10)14-5-3-1-2-4-6(5)15(11,12)13/h1-4H,(H2,11,12,13)
SMILES C1=CC=C(C(=C1)OC(F)(F)F)S(=O)(=O)N
IUPAC Name 2-(trifluoromethoxy)benzenesulfonamide
Molecular Weight (g/mol) 241.19
Formula Weight 241.002
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.