missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, TRC

Product Code. 30452088
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30452088

Brand: Toronto Research Chemicals TRCT79069525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Trifluoromethoxy)aniline
CAS 1535-75-7
Molecular Formula C7 H6 F3 N O
Quantity 25 g
Synonym Benzenamine','2-(trifluoromethoxy)-, o-Anisidine','α,α,α-trifluoro- (6CI,7CI,8CI), 2-(Trifluoromethoxy)benzenamine, 2-(Trifluoromethyloxy)aniline, 2-Trifluoromethoxyaniline, 2-Trifluoromethoxyphenylamine, o-(Trifluoromethoxy)aniline, o-(Trifluoromethoxy)phenylamine
InChI Formula InChI=1S/C7H6F3NO/c8-7(9,10)12-6-4-2-1-3-5(6)11/h1-4H,11H2
SMILES Nc1ccccc1OC(F)(F)F
IUPAC Name 2-(trifluoromethoxy)aniline
Molecular Weight (g/mol) 177.12
Formula Weight 177.0401
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.