missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10504704
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
This item is not returnable. View return policy

Product Code. 10504704

Brand: Thermo Scientific Maybridge SB01054FA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00035959
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CF3OC6H4NH2
Quantity 100 g
Formula Weight 177.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.