missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoroacetyl)thiophene, 98%

Product Code. 11486780
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11486780 5 g 5g
11476780 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11486780

Brand: Thermo Scientific Alfa Aesar L12585.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 651-70-7 | C6H3F3OS | 180.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-70-7
Molecular Formula C6H3F3OS
Molecular Weight (g/mol) 180.144
MDL Number MFCD00041427
InChI Key CZYKJGCKVBXLGF-UHFFFAOYSA-N
Synonym 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene
PubChem CID 136464
IUPAC Name 2,2,2-trifluoro-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)C(F)(F)F

Specifications

Density 1.403
Boiling Point 163°C to 165°C
Flash Point 58°C (136°F)
Refractive Index 1.4845
Quantity 5 g
UN Number UN1993
Beilstein 1309208
Formula Weight 180.15
Percent Purity 98%
Chemical Name or Material 2-(Trifluoroacetyl)thiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.