missing translation for 'onlineSavingsMsg'
Learn More

2-Tridecanone, 98+%

Product Code. 11429346
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429346 25 g 25g
11439346 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429346 Supplier Thermo Scientific Alfa Aesar Supplier No. A19042.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-08-8 | C13H26O | 198.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-08-8
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00008968
InChI Key CYIFVRUOHKNECG-UHFFFAOYSA-N
Synonym 2-tridecanone, methyl undecyl ketone, mathyl undecyl kepoje, hendecyl methyl ketone, 2-tridecankje, tridecanone-2, methyl n-undecyl ketone, tridecanone, 2-tridecanone natural, unii-5q35vhx26k
PubChem CID 11622
ChEBI CHEBI:77928
IUPAC Name tridecan-2-one
SMILES CCCCCCCCCCCC(=O)C

Specifications

Melting Point 29°C to 31°C
Boiling Point 133°C to 134°C (10 mmHg)
Flash Point 122°C (251°F)
Odor Characteristic
Quantity 25 g
UN Number UN3082
Beilstein 1757402
Formula Weight 198.35
Percent Purity ≥98%
Chemical Name or Material 2-Tridecanone
compliance-icons
Product Identifier
  • 2-Tridecanone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Methyl undecyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.