missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trichloroacetyl)pyrrole, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-(Trichloroacetyl)pyrrole |
| CAS | 35302-72-8 |
| Molecular Formula | C6H4Cl3NO |
| Quantity | 1 g |
| Synonym | 2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone, 2-(Trichloroacetyl)-1H-pyrrole, NSC 272669, Pyrrol-2-yl Trichloromethyl Ketone |
| InChI Formula | InChI=1S/C6H4Cl3NO/c7-6(8,9)5(11)4-2-1-3-10-4/h1-3,10H |
| SMILES | ClC(Cl)(Cl)C(=O)c1ccc[nH]1 |
| IUPAC Name | 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone |
| Molecular Weight (g/mol) | 212.46 |
| Formula Weight | 210.9358 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction