missing translation for 'onlineSavingsMsg'
Learn More

2-(Trichloroacetyl)pyrrole, 99+%

Product Code. 15410559
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410559 25 g 25g
15400559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410559 Supplier Thermo Scientific Alfa Aesar Supplier No. H63718.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35302-72-8 | C6H4Cl3NO | 212.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35302-72-8
Molecular Formula C6H4Cl3NO
Molecular Weight (g/mol) 212.454
MDL Number MFCD00128757
InChI Key BBFDGMDENAEMKF-UHFFFAOYSA-N
Synonym 2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole
PubChem CID 321487
IUPAC Name 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
SMILES C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl

Specifications

Melting Point 72°C to 74°C
Color Gray
Quantity 25 g
Beilstein 135479
Formula Weight 212.46
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-(Trichloroacetyl)pyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.