missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)oxazole, 95%

Product Code. 15422258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422258 5 g 5g
15432258 250 mg 250mg
15412258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422258 Supplier Thermo Scientific Alfa Aesar Supplier No. H55870.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 145214-05-7 | C15H29NOSn | 358.113 g/mol

2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145214-05-7
Molecular Formula C15H29NOSn
Molecular Weight (g/mol) 358.113
MDL Number MFCD06798076
InChI Key YOWGRWHKDCHINP-UHFFFAOYSA-N
Synonym 2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole
PubChem CID 15486183
IUPAC Name tributyl(1,3-oxazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CO1

Specifications

Color Colorless
Density 1.71 g/mL
Boiling Point 108°C to 110°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.493
Quantity 5 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 358.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)oxazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.