missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxotetrahydro-1,3-oxazole, 98%, Thermo Scientific™

Product Code. 10697251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697251 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10697251 Supplier Thermo Scientific Acros Supplier No. 304135000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5840-81-3
CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Boiling Point 85.0°C to 91.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C3H5NOS
MDL Number MFCD00125763
Quantity 500 mg
Synonym oxazolidine-2-thione, 2-oxazolidinethione, oxazolidinethione, 2-thiooxazolidone, 2-mercapto-2-oxazoline, 2-thioxotetrahydro-1,3-oxazole, unii-6smj3cuw4c, 1,3-oxazolane-2-thione, 6smj3cuw4c, 4,5-dihydro-1,3-oxazole-2-thiol
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
InChI Key UMURLIQHQSKULR-UHFFFAOYSA-N
SMILES C1COC(=S)N1
IUPAC Name 1,3-oxazolidine-2-thione
Molecular Weight (g/mol) 103.14
PubChem CID 2733223
Formula Weight 103.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thioxotetrahydro-1, 3-oxazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.