Learn More
2-Thiouracil, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• 2-Thiouracil (Thiouracil or 4-Hydroxy-2-mercaptopyrimidine) is a nucleotide analog with a C-2 thioxo substitution
• 2-Thiouracil is a melanocyte suppressor and thyroid inhibitor
Applications
• 2-Thiouracil can be used to inhibit nitric oxide synthase and suppresses nitric oxide signaling in vitro
• 2-Thiouracil can be used for labeling of RNA in vitro
• 2-Thiouracil is intended for research applications only
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | >300.0°C |
Color | White to Cream |
Loss on Drying | 0.5% max. (105°C, 3 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Plastic bottle |
Quantity | 500 g |
Beilstein | 24, 323 |
Show More |
RUO – Research Use Only