missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopyridone, TRC

Product Code. 30456102
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456102 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456102 Supplier Toronto Research Chemicals Supplier No. TRCT3682905G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Thiopyridone
CAS 2637-34-5
Molecular Formula C5H5NS
Quantity 5 g
Synonym 2(1H)-Pyridinethione, 1H-Pyridine-2-thione, 2-Mercaptopyridine, 2-Pyridinethiol, Alrithiol-2, NSC 41337, WR 608
InChI Formula InChI=1S/C5H5NS/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)
SMILES Sc1ccccn1
IUPAC Name pyridine-2-thiol
Molecular Weight (g/mol) 111.16
Formula Weight 111.0143
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.