missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethylamine, 94%

Product Code. 11439213
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439213 5 g 5g
11449213 25 g 25g
11459213 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11700.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27757-85-3 | C5H7NS | 113.178 g/mol

2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.

Solubility
Easily soluble in ethanol and Chloroform.

Notes
Air Sensitive. Store under dry inert gas. Store away from air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27757-85-3
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005460
InChI Key FKKJJPMGAWGYPN-UHFFFAOYSA-N
Synonym 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
PubChem CID 34005
IUPAC Name thiophen-2-ylmethanamine
SMILES C1=CSC(=C1)CN

Specifications

Density 1.1
Boiling Point 85°C to 88°C (14 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.567
Quantity 5 g
UN Number UN2735
Beilstein 106296
Sensitivity Air Sensitive
Solubility Information Easily soluble in ethanol and Chloroform.
Formula Weight 113.19
Percent Purity 97%
Chemical Name or Material 2-Thiophenemethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.