Learn More
2-Thiophenemethylamine, 94%
Description
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
Solubility
Easily soluble in ethanol and Chloroform.
Notes
Air Sensitive. Store under dry inert gas. Store away from air.
Specifications
Specifications
| Density | 1.1 |
| Boiling Point | 85°C to 88°C (14 mmHg) |
| Flash Point | 73°C (163°F) |
| Refractive Index | 1.567 |
| Quantity | 25 g |
| UN Number | UN2735 |
| Beilstein | 106296 |
| Sensitivity | Air Sensitive |
| Solubility Information | Easily soluble in ethanol and Chloroform. |
| Formula Weight | 113.19 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.