missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals

Product Code. 10216360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216360 100 g Glass bottle 100g
10328680 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216360 Supplier Thermo Scientific Acros Supplier No. 205571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27757-85-3
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.18
MDL Number MFCD00005460
InChI Key FKKJJPMGAWGYPN-UHFFFAOYSA-N
Synonym 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
PubChem CID 34005
IUPAC Name thiophen-2-ylmethanamine
SMILES C1=CSC(=C1)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 95.0°C to 99.0°C (28.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5650 to 1.5680
Quantity 100 g
Beilstein 18, IV, 7096
Specific Gravity 1.1
Formula Weight 113.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethylamine, 90-96%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenemethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.