missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethanol, 99+%

Product Code. 10215430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215430 25 g Glass bottle 25g
10742705 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215430 Supplier Thermo Scientific Acros Supplier No. 138860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-72-6 | C5H6OS | 114.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-72-6
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00005454
InChI Key ZPHGMBGIFODUMF-UHFFFAOYSA-N
Synonym 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
PubChem CID 69467
SMILES OCC1=CC=CS1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 206.0°C to 207.0°C (761.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5622 to 1.5642
Quantity 25 g
Beilstein 17, 113
Specific Gravity 0.9
Solubility Information Solubility in water: 40g/L (20°C).
IUPAC Name (thiophen-2-yl)methanol
Formula Weight 114.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.