missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneethylamine, 98%

Product Code. 11444313
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444313 25 g 25g
11424313 1 g 1g
11434313 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444313 Supplier Thermo Scientific Alfa Aesar Supplier No. A10920.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30433-91-1 | C6H9NS | 127.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30433-91-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00051495
InChI Key HVLUYXIJZLDNIS-UHFFFAOYSA-N
Synonym thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
PubChem CID 116521
IUPAC Name 2-thiophen-2-ylethanamine
SMILES C1=CSC(=C1)CCN

Specifications

Density 1.087
Boiling Point 201°C
Flash Point 87°C (188°F)
Refractive Index 1.551
Quantity 25 g
UN Number UN2735
Beilstein 106962
Sensitivity Air sensitive
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material 2-Thiopheneethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.