missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneethanol, 98%

Product Code. 11497154
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497154 100 g 100g
11477154 5 g 5g
11487154 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497154 Supplier Thermo Scientific Alfa Aesar Supplier No. B21562.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5402-55-1 | C6H8OS | 128.19 g/mol

2-Thiopheneethanol, is used as the antithrombotics thiophene chlorine match piperidine intermediates. It is also used as a ticlopidine Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiopheneethanol, is used as the antithrombotics thiophene chlorine match piperidine intermediates. It is also used as a ticlopidine Intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5402-55-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00005462
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
PubChem CID 79400
IUPAC Name 2-thiophen-2-ylethanol
SMILES OCCC1=CC=CS1

Specifications

Density 1.15
Boiling Point 107°C to 108°C (12 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.551
Quantity 100 g
Beilstein 106985
Solubility Information Slightly soluble in water.
Formula Weight 128.19
Percent Purity 98%
Chemical Name or Material 2-Thiopheneethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.