missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetic Acid, TRC

Product Code. 30455675
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455675 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455675 Supplier Toronto Research Chemicals Supplier No. TRCT3447951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Thiopheneacetic Acid
CAS 1918-77-0
Molecular Formula C6H6O2S
Quantity 1 g
Synonym 2-(2-Thienyl)acetic Acid, 2-(Thien-2-yl)acetic Acid, 2-(Thiophen-2-yl)acetic Acid, 2-Thienylacetic Acid, 2-Thienylethanoic Acid, Thiophen-2-ylacetic Acid
InChI Formula InChI=1S/C6H6O2S/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2,(H,7,8)
SMILES O=C(O)CC1=CC=CS1
IUPAC Name 2-thiophen-2-ylacetic acid
Molecular Weight (g/mol) 142.18
Formula Weight 142.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.