missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetic acid, 98%

Product Code. 10232652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10232652 5 g Glass Bottle 5g
10307610 25 g Plastic bottle 25g
10427570 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10232652 Supplier Thermo Scientific Acros Supplier No. 187730050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1918-77-0 | C6H6O2S | 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-77-0
MDL Number MFCD00005458
InChI Key SMJRBWINMFUUDS-UHFFFAOYSA-N
Synonym 2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
PubChem CID 15970
ChEBI CHEBI:45807
IUPAC Name 2-thiophen-2-ylacetic acid
SMILES C1=CSC(=C1)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 61°C to 65°C
Color Brown
Boiling Point 160°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C6H6O2S
Quantity 5 g
Beilstein 18, 293
Molecular Weight (g/mol) 142.17
Formula Weight 142.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiopheneacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiopheneacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.