Learn More
2-Thiopheneacetic acid, 98%
Description
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 61°C to 65°C |
| Boiling Point | 160°C (22 mmHg) |
| Quantity | 500 g |
| UN Number | UN3261 |
| Beilstein | 114551 |
| Solubility Information | Soluble in water,ethanol,ether and carbon tetrachloride. |
| Formula Weight | 142.18 |
| Percent Purity | 98% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Thiopheneacetic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.